missing translation for 'onlineSavingsMsg'
Learn More

Famotidine, 98+%, Thermo Scientific™

Product Code. 15415429
1g
Click to view available options
Quantity:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

A competitive histamine H2 receptor antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water and methanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76824-35-6
Molecular Formula C8H15N7O2S3
Molecular Weight (g/mol) 337.435
MDL Number MFCD00079297
InChI Key XUFQPHANEAPEMJ-UHFFFAOYSA-N
Synonym famotidine, pepcid ac, apogastine, amfamox, antodine, bestidine, digervin, dispromil, famodil, gastridin
PubChem CID 5702160
ChEBI CHEBI:4975
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide
SMILES C1=C(N=C(S1)N=C(N)N)CSCCC(=NS(=O)(=O)N)N

Specifications

Quantity 1g
Merck Index 14,3930
Solubility Information Soluble in water and methanol
Formula Weight 337.45
Percent Purity ≥98%
Chemical Name or Material Famotidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Famotidine, 98+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.