Learn More
Hellma™ Far UV Quartz Matched Pairs Semi-Micro Cells
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Far UV Quartz Matched Pairs Semi-Micro Cuvettes are manufactured from quartz, which is chemically resistant and maintains its shape, even at high temperatures up to approximately 1000°C.
Brand: Hellma™ 115-10-40E
This item is not returnable.
View return policy
Description
- High UV transmission
- Low thermal expansion
Specifications
Matched Pairs Semi-micro Cell | |
Far UV Quartz | |
200 to 2500 nm |
0.4 mL | |
10 mm |