missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FARSB Monoclonal Antibody (OTI4B3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA526809
317.10 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.
This item is not returnable.
View return policy
Description
This gene encodes a highly conserved enzyme that belongs to the aminoacyl-tRNA synthetase class IIc subfamily. This enzyme comprises the regulatory beta subunits that form a tetramer with two catalytic alpha subunits. In the presence of ATP, this tetramer is responsible for attaching L-phenylalanine to the terminal adenosine of the appropriate tRNA. A pseudogene located on chromosome 10 has been identified.
Specifications
FARSB | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NSD9 | |
Farsb | |
Full length human recombinant protein of FARSB produced in HEK293T cell | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4B3 | |
Unconjugated | |
Farsb | |
C76708; Farsa; FARSB; Farsl; Farslb; Frsb; HSPC173; ph; PheHB; phenylalanine tRNA ligase 1, beta, cytoplasmic; phenylalanine tRNA synthetase alpha; phenylalanine-tRNA ligase beta chain; phenylalanine--tRNA ligase beta chain; phenylalanine--tRNA ligase beta subunit; phenylalanine-tRNA synthetase-like, beta subunit; phenylalanyl-tRNA synthetase beta chain; phenylalanyl-tRNA synthetase beta subunit; phenylalanyl-tRNA synthetase beta-subunit; phenylalanyl-tRNA synthetase, beta subunit; phenylalanyl-tRNA synthetase-like, beta subunit; PheRS; PheRS alpha | |
Mouse | |
Affinity Chromatography | |
RUO | |
10056 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FARSB Monoclonal Antibody (OTI4B3), Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction