missing translation for 'onlineSavingsMsg'
Learn More

Fasentin, Tocris Bioscience™

Product Code. 30001000
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GLUT1/GLUT4 inhibitor; also Fas-sensitizer

GLUT1 and GLUT4 inhibitor (IC50 = 68 μM). Preferentially inhibits GLUT4 over GLUT1. Sensitizes Fas receptor in a range of tumor cell lines (IC50 = 20 μM) by modulating the extrinsic apoptotic pathway downstream of TNF receptors but upstream of effector caspases.

Chemical Identifiers

CAS 392721-37-8
Molecular Formula C11H9ClF3NO2
Molecular Weight (g/mol) 279.64
MDL Number MFCD01001285
InChI Key GNYIJZMBLZXJEJ-UHFFFAOYSA-N
Synonym Fasentin
PubChem CID 879520
IUPAC Name N-[4-chloro-3-(trifluoromethyl)phenyl]-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC(=C(Cl)C=C1)C(F)(F)F

Specifications

Linear Formula C11H9ClF3NO2
Quantity 10 mg
Solubility Information Soluble to 100mM in DMSO and to 100mM in ethanol
Formula Weight 279.64
Percent Purity >98%
Physical Form Solid
Chemical Name or Material N-(4-Chloro-3-(trifluoromethyl)phenyl)-3-oxobutanamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'