missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FASN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522061
This item is not returnable.
View return policy
Description
Recommended positive controls: Molt-4, mouse brain, PC-12, Rat2, 293T. Predicted reactivity: Mouse (89%), Rat (89%), Pig (83%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Fatty acid synthase is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-alpha (ER-alpha), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha.Specifications
FASN | |
Polyclonal | |
Unconjugated | |
FASN | |
[Acyl-carrier-protein] S-acetyltransferase; [Acyl-carrier-protein] S-malonyltransferase; 3-hydroxyacyl-[acyl-carrier-protein] dehydratase; 3-oxoacyl-[acyl-carrier-protein] reductase; 3-oxoacyl-[acyl-carrier-protein] synthase; A630082H08Rik; Enoyl-[acyl-carrier-protein] reductase; FAS; FASN; Fatty acid synthase; fatty acid synthase (EC 3.2.1.23); MGC14367; MGC15706; OA-519; Oleoyl-[acyl-carrier-protein] hydrolase; SDR27X1; short chain dehydrogenase/reductase family 27X, member 1; thioesterase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14104, 2194, 50671 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P12785, P19096, P49327 | |
FASN | |
Recombinant fragment corresponding to a region within amino acids 7 and 287 of Fatty Acid Synthase (Uniprot ID#P49327) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |