Learn More
Epredia™ Fast Flex™ Holding Solution
Employ a fixative designed for use in the grossing room with Fast Flex™ Holding Solution.
Brand:  Epredia™ 9800
Code : A0
Additional Details : CAS Number : 50-00-0 Weight : 0.01000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
 View more versions of this product
Product Code. 12559946
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Place cassettes in holding solution for at least 20 minutes prior to loading on the processor
  • Compatible with 10% Neutral Buffered Formalin and other fixatives
Specifications
Specifications
2.5 gal.