missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Fast Flex™ Holding Solution

Employ a fixative designed for use in the grossing room with Fast Flex™ Holding Solution.

Brand:  Epredia™ 9800-1

Code : A0

Additional Details : Weight : 0.01000kg Transport : UN number : 1170 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 12569946

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Place cassettes in holding solution for at least 20 minutes prior to loading on the processor
  • Compatible with 10% Neutral Buffered Formalin and other fixatives
Specifications

Specifications

Holding Solution
5L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • fast flex hlding solution

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1A
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H350-May cause cancer.
  • H370-Causes damage to organs.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P363-Wash contaminated clothing before reuse.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P370+P378-In case of fire: Use CO2, dry chemical orfoam to extinguish.
  • P405-Store locked up.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, formaldehyde, methanol, IPA

SDS
Documents

Documents

Certificates
Special Offers

Special Offers