missing translation for 'onlineSavingsMsg'
Learn More

Fast Garnet GBC Base 97%, Thermo Scientific Chemicals

Product Code. 10501981 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 10501981

Brand: Thermo Scientific Acros 153260250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-56-3
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.295
MDL Number MFCD00007733
InChI Key PFRYFZZSECNQOL-UHFFFAOYSA-N
Synonym 2-Aminoazotoluene, 4'-Amino-2, 3'-dimethylazobenzene, C.I. 11160, Solvent Yellow 3
PubChem CID 7340
IUPAC Name 2-methyl-4-[(2-methylphenyl)diazenyl]aniline
SMILES CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C

Specifications

Assay Percent Range 97%
Linear Formula NH2C6H3(CH3)N=NC6H4CH3
Beilstein 16, 344
Merck Index 15, 415
Infrared Spectrum Authentic
Formula Weight 225.29
Percent Purity ≥96.0%
Packaging Glass bottle
UV Spectrometry Complies
Color Brown/Red
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Fast Garnet GBC base, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Fast Garnet GBC base
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Respiratory sensitiser Category 1
Hazard Statement
  • H350-May cause cancer.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.