Learn More
Fast Garnet GBC Base 97%, Thermo Scientific Chemicals
£28.90
Chemical Identifiers
CAS | 97-56-3 |
---|---|
Molecular Formula | C14H15N3 |
Molecular Weight (g/mol) | 225.295 |
MDL Number | MFCD00007733 |
InChI Key | PFRYFZZSECNQOL-UHFFFAOYSA-N |
Synonym | 2-Aminoazotoluene, 4'-Amino-2, 3'-dimethylazobenzene, C.I. 11160, Solvent Yellow 3 |
PubChem CID | 7340 |
IUPAC Name | 2-methyl-4-[(2-methylphenyl)diazenyl]aniline |
SMILES | CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97-56-3 | |
225.295 | |
PFRYFZZSECNQOL-UHFFFAOYSA-N | |
7340 | |
CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C |
C14H15N3 | |
MFCD00007733 | |
2-Aminoazotoluene, 4'-Amino-2, 3'-dimethylazobenzene, C.I. 11160, Solvent Yellow 3 | |
2-methyl-4-[(2-methylphenyl)diazenyl]aniline |
Specifications
97-56-3 | |
C14H15N3 | |
MFCD00007733 | |
15, 415 | |
PFRYFZZSECNQOL-UHFFFAOYSA-N | |
CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C | |
225.295 | |
225.29 | |
Glass bottle | |
Brown/Red | |
Fast Garnet GBC base, 97% |
97% | |
NH2C6H3(CH3)N=NC6H4CH3 | |
16, 344 | |
2-Aminoazotoluene, 4'-Amino-2, 3'-dimethylazobenzene, C.I. 11160, Solvent Yellow 3 | |
Authentic | |
2-methyl-4-[(2-methylphenyl)diazenyl]aniline | |
7340 | |
≥96.0% | |
Complies | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
May cause cancer.
GHS P Statement:
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 202-591-2
missing translation for 'rtecsNumber' : XU8800000
missing translation for 'tsca' : TSCA
- Fast Garnet GBC base
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
- Respiratory sensitiser Category 1
Hazard Statement
- H350-May cause cancer.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only