Learn More
Thermo Scientific™ FastDigest SmaI
Shop All Thermo Scientific ProductsDescription
5' CCC ↓ GGG 3'
3' GGG ↑ CCC 5'
FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.
The universal buffer allows rapid single-, double- or multiple-DNA digestion within 5-15 minutes, eliminating any need for buffer change or subsequent DNA clean-up steps. DNA modifying enzymes, such as Klenow Fragment, T4 DNA ligase, alkaline phosphatases, and T4 DNA polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.
For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.
Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.
Features
- 100% activity of all FastDigest enzymes in the universal buffer
- 100% buffer compatibility with downstream applications
- Complete digestion in 5–15 minutes
- Direct loading on gels
- No star activity
- 176 FastDigest enzymes available
Applications
- Molecular cloning
- Restriction site mapping
- Genotyping
- Southern Blot
- Restriction fragment length polymorphism (RFLP)
- SNP analysis
Notes
- The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer.
- For applications that require product analysis by fluorescence excitation (e.g., concentration measurements in UV light), the colorless FastDigest Buffer is recommended.
- For methylation sensitivity, refer to product specifications.
Specifications
Specifications
| Content And Storage | FastDigest SmaI (200 μL), 10X FastDigest Buffer (1.0 mL), 10X FastDigest Green Buffer (1.0 mL), Store at –20°C. |
| Methylation Sensitivity | CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive |
| Enzyme | Sma I |
| Compatible Buffer | 10X FastDigest Buffer/FastDigest Green Buffer |
| Sensitive to Heat Inactivation | Yes |
| Optimal Reaction Temperature | 37°C |
| Type IIS RE | No |
| Quantity | 200 μL (200 Reactions) |
| Product Type | Restriction Enzyme |
| Research Category | Traditional Cloning |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.