missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals FastDNA™ SPIN Kit for Soil

FastDNA™ SPIN Kit for Soil is used for rapid isolation of PCR-ready genomic DNA from any environmental sample.

Brand:  MP Biomedicals 116560200

545.29 GBP valid until 2024-12-31
Use promo code "21949" to get your promotional price.



 View more versions of this product

Product Code. 11492400

  • £720.00 / 50 preps

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The FastDNA™ SPIN Kit for Soil quickly and efficiently isolates bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples.
  • Soil is processed by a FastPrep™ instrument and Lysing Matrix E tubes designed to efficiently lyse all microorganisms.
  • The released DNA is purified by a silica-based spin filter method and is suitable for PCR analysis and other downstream applications.
  • Complete lysis of all organisms, including historically difficult sources such as eubacterial spores and endospores, gram positive bacteria, yeast, algae, nematodes and fungi is achieved within seconds using a FastPrep instrument and Lysing Matrix E tubes.
  • The FastDNA Spin Kit for Soil delivers the highest DNA yield from any environmental sample including soil, sediments, sludge, compost, manure, rhizosphere or waste water.
  • The FastDNA Spin Kit for Soil contains Lysing Matrix tubes and all the reagents needed to extract DNA in a process that efficiently removes humic acids and other PCR inhibitors.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 mL
Kit
Soil, Feces, Sludge, Wastewater, Environmental, Compost, Manure, Rhizosphere
Isolate bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples, including sediment, sludge, compost, manure, rhizosphere, waste water and more
50 Preps
Isolate bacterial, fungal, plant, and animal genomic DNA
30 min.
Up to 500 mg per prep
Glass Milk
FastPrep™ Instrument
Lysing Matrix E
Nucleic Acid Isolation, Mini-prep
DNA
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • FastDNA SPIN Kit for Soil

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 A
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P264-Wash thoroughly after handling.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P330-Rinse mouth.

Supplemental information
  • MIXTURE LIST-Contains : Sodium Dodecyl Sulfate, Polyvinylpyrrolidone (PVP 40), Guanidine Thiocyanate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.