missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Fastening Clip
Winkler pattern. For use with oxygen flasks.
Brand: BRAND™ 386148
This item is not returnable.
View return policy
Description
- Metal construction
- Winkler pattern
- Fits oxygen flasks with 250 to 300mL capacity
Specifications
1 Ea. | |
Oxygen flasks | |
Winkler pattern | |
250 to 300 mL |