missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Checkers Fastlane™ 1 Channel 1.5'' Drop Over Cable Protector
Fastlane lightweight drop-over cable protectors are made from durable polyurethane and feature heavy-duty, patented L-Connectors for easily attaching additional protectors to extend to any desired length.
Brand: Checkers PFLS0002OR
Additional Details : Weight : 1.40000kg
Description
- Fastlane lightweight drop-over cable protectors are made from durable polyurethane and feature heavy-duty, patented L-Connectors for easily attaching additional protectors to extend to any desired length
- Patented 5-bar tread plate surface provides maximum traction for carts and pedestrian traffic
- These low profile protectors fit multiple cable sizes and are compact for easy setup, disassembly, and storage
Specifications
1.5 in. | |
Cable Protector | |
Polyethylene |
Orange | |
91.4 cm | |
3.8 cm |