missing translation for 'onlineSavingsMsg'
Learn More

Checkers Fastlane™ 1 Channel 1.5 in. Drop Over Cable Protector

Fastlane lightweight drop-over cable protectors are made from durable polyurethane and feature heavy-duty, patented L-Connectors for easily attaching additional protectors to extend to any desired length.

Brand:  Checkers PFLS0001BL

 View more versions of this product

Product Code. 17782398

  • £48.50 / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Fastlane lightweight drop-over cable protectors are made from durable polyurethane and feature heavy-duty, patented L-Connectors for easily attaching additional protectors to extend to any desired length
  • Patented 5-bar tread plate surface provides maximum traction for carts and pedestrian traffic
  • These low profile protectors fit multiple cable sizes and are compact for easy setup, disassembly, and storage
Specifications

Specifications

1.5 in.
91.4 cm
Polyethylene
3.8 cm
Black
Cable Protector
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.