Learn More
Thermo Scientific™ FastRuler Ultra Low Range DNA Ladder, Ready-to-Use, 10-200 bp PROMO
Perform fast sizing and approximate quantification of dsDNA in high throughput gels and conventional agarose gels with Thermo Scientific™ FastRuler™ DNA Ladders.
Brand: Thermo Scientific™ SM1233-RMD123
Description
Highlights
- Fast separation (8 to 14 min)
- Short separation distance (10 to 20 mm)
- Sharp bands, easy-to-remember fragment sizes and quantities
- Ready-to-use -premixed with loading dye
- Supplied with loading dye solution for sample DNA
Specifications
Orange | |
DNA Molecular Weight Marker/Ladder | |
1 mL |
6X Loading Dye | |
Liquid |
This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount of the product in internal research conducted by the buyer. The buyer cannot use this product or materials made by the employment of this product for Commercial Purposes. Commercial Purposes means any activity for consideration and may include, but is not limited to: (1) use of the product in manufacturing; (2) use of the product to provide a service, information, or data; (3) use of the product for therapeutic, diagnostic or prophylactic purposes; or (4) resale of the product, whether or not the product is resold for use in research. Further information on purchasing licenses under the above patents may be obtained by contacting the Business Development Department, Invitrogen Corporation, 5791 Van Allen Way, Carlsbad, CA 92008.