missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO33, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00254170-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MLLFLSVPQPRPPGARTRAGAARVARWRRLRLQQLRRLRGLLRVLRGRPGAGSRRRGRMALCGQAAGAASLPSELIVHIFSFLPAPDRLRASASCSHWRECLFYPALWPQLRICLRVSPAEQPRLEFLMRKCGWFVRELRVEFAAENYLSGGGPGDGGGADTGTGGEEVEALQLSARWLEVLRTYLELVLCVLVSIRNNRNLQKFSLFGDISVLQQQGSLSNTYLSKVDPDGKKIKQIQQLFEEILSNSRQLKWLSCGFMLEIVTPTSLSSLSNAVANTMEHLSLLDNNIPGNSTLITAVELERFVNLHSLALDFCDFTAEMARVLTDSNHVPLQRLSLLVHNVSVMHKSLDNMPNDEHWKALSRKSTSFRVYIMAFDIKSEDMLKILKPSIPLERIHFDSYITCVSGAIVDLISRQYDKFLTHFILMNDVIDTSGFPDLSDNRNEDPLVLLAWRCTKLSLLAIHGYTVWAHNLIAIARLRGSDLKVLEVTEESIDFDQGELADQDVDPVHNLIEQVSLGLGQPWHAVMDIESLSVFTEPNRHFYREMQSFSEDISpecifications
FBXO33 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
NM_203301.1 | |
FBXO33 | |
FBXO33 (NP_976046.1, 1 a.a. ∼ 555 a.a) full-length human protein. | |
RUO | |
254170 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
F-box protein 33 | |
Fbx33/c14_5247 | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction