missing translation for 'onlineSavingsMsg'
Learn More

FCCP, Tocris Bioscience™

Product Code. 18742724
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Oxidative phosphorylation uncoupler

A very potent uncoupler of oxidative phosphorylation in mitochondria.

Chemical Identifiers

CAS 370-86-5
Molecular Formula C10H5F3N4O
Molecular Weight (g/mol) 254.172
InChI Key BMZRVOVNUMQTIN-UHFFFAOYSA-N
Synonym fccp, carbonyl cyanide p-trifluoromethoxyphenylhydrazone, carbonyl cyanide 4-trifluoromethoxy phenylhydrazone, 2-2-4-trifluoromethoxy phenyl hydrazono malononitrile, propanedinitrile, 4-trifluoromethoxy phenyl hydrazono, 1-cyano-n-4-trifluoromethoxy phenyl methanecarbohydrazonoyl cyanide, carbonyl cyanide 4-trifluoromethoxyphenylhydrazone, carbonyl cyanide, 4-trifluoromethoxyphenylhydrazone, 4-trifluoromethoxy phenyl hydrazono malononitrile, mesoxalonitrile, p-trifluoromethoxy phenyl hydrazone
PubChem CID 3330
ChEBI CHEBI:75458
IUPAC Name 2-[[4-(trifluoromethoxy)phenyl]hydrazinylidene]propanedinitrile
SMILES C1=CC(=CC=C1NN=C(C#N)C#N)OC(F)(F)F

Specifications

Quantity 10 mg
Formula Weight 254.17
Percent Purity >99%
Chemical Name or Material FCCP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
FCCP, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.