missing translation for 'onlineSavingsMsg'
Learn More
Febantel, Thermo Scientific™ Product Code.: 17322501

Febantel, Thermo Scientific™

Product Code. 17322501
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 17322501

Brand: Thermo Scientific Acros 468780050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 58306-30-2
Molecular Formula C20H22N4O6S
Molecular Weight (g/mol) 446.48
MDL Number 01738527
InChI Key HMCCXLBXIJMERM-UHFFFAOYSA-N
IUPAC Name methyl N-{[(methoxycarbonyl)imino]({[2-(2-methoxyacetamido)-4-(phenylsulfanyl)phenyl]amino})methyl}carbamate
SMILES COCC(=O)NC1=CC(SC2=CC=CC=C2)=CC=C1NC(NC(=O)OC)=NC(=O)OC

Specifications

Packaging Glass Bottle
Quantity 5 g
Certificate of Analysis Available
Physical Form Powder/Crystals
Merck Index 3973
Color White to Light Yellow
Melting Point 129°C to 130°C
Solubility Insoluble in water
Formula Weight 446.48
Chemical Name or Material Febantel
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Febantel, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.