Learn More
Fehling's reagent II for sugars, For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline, Honeywell Fluka™
for determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline
Brand: Honeywell Fluka 36019-6X500ML
Code : NEW
Additional Details : Weight : 3.72000kg Transport : UN number : 1824 Chem class : 8 Pack group : II
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Fehling’s reagent II for sugars | |
MFCD00012467 | |
1.23-1.25g/mL at 20°C |
1310-73-2,304-59-6,7732-18-5 | |
UN1824 | |
6 x 500 mL |