missing translation for 'onlineSavingsMsg'
Learn More

Fehling's reagent II for sugars, For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline, Honeywell Fluka™

for determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline

Brand:  Honeywell-Fluka 36019-1L

86.88 GBP valid until 2025-12-31
Use promo code "24376" to get your promotional price.



 View more versions of this product

Product Code. 15635040

  • £91.45 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fehling’s reagent II for sugars
MFCD00012467
1.23-1.25g/mL at 20°C
1310-73-2, 304-59-6, 7732-18-5
UN1824
1 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Fehling

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.