Learn More
Fehling's reagent II for sugars, For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline, Honeywell Fluka™
for determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline
Brand: Honeywell Fluka 36019-4X2.5L
Code : NEW
Additional Details : Weight : 12.40000kg Transport : UN number : 1824 Chem class : 8 Pack group : II
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Fehling’s reagent II for sugars | |
1310-73-2,304-59-6,7732-18-5 | |
≥10.0000% | |
MFCD00012467 | |
1.23-1.25 g/mL at 20 °C | |
Reagent Ph. Eur. | |
4 x 2.5 L |
For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline | |
100°C | |
<30.0000% | |
UN1824 | |
Potassium sodium tartrate solution, alkaline, Alkaline potassium sodium tartrate | |
Poly Bottle |