missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fehling's reagent II for sugars, For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline, Honeywell Fluka™
for determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline
Brand: Honeywell-Fluka 36019-6X1L
521.14 GBP valid until 2025-12-31
Use promo code "24376" to get your promotional price.
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Fehling’s reagent II for sugars | |
1310-73-2, 304-59-6, 7732-18-5 | |
≥70.0000%, ≥10.0000% | |
MFCD00012467 | |
1.23-1.25 g/mL at 20 °C | |
Reagent Ph. Eur. | |
6 x 1 L |
For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline | |
100°C | |
<90.0000%, <30.0000% | |
UN1824 | |
Potassium sodium tartrate solution, alkaline, Alkaline potassium sodium tartrate | |
Poly Bottle |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Fehling
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction