missing translation for 'onlineSavingsMsg'
Learn More

Fehlings No.2 Indicator Solution, Reagecon
BIOPHARMA

High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.

Brand:  Reagecon FS0102


Product Code. 11573192

  • £111.00 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • For manual titrations; indicator titrant or analyte change color and this color change are by far the most important method of end point detection.
  • This high-quality indicator solution facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications

Specifications

HDPE Bottle GL45
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • FEHLINGS SOLUTION No.2

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers