missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Feline IL-10 DuoSet ELISA
Feline IL-10 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DY736
Additional Details : Weight : 0.09700kg
Description
The Feline IL-10 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies feline IL-10.Specifications
Biotin | |
ELISA | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF |
125.00 to 8000 pg/mL | |
493683 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |