Learn More
Masterflex™ Female Luer to 1/8 in. Low-profile Semi-rigid Tubing Barb
Ideal for a variety of critical applications, including syringes or small tubing
Brand: Masterflex™ 45508-16
Code : NEW
Additional Details : Weight : 0.00907kg
Description
- Cole-Parmer offers a variety of barbed luer adapters to meet your application's needs.
- Standard hose barb for use with flexible tubing.
- Smooth bore hose barb to reduce flow turbulence and risk of line contamination.
- Wide-bore hose barb for enhanced fluid path flow.
- Low-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene.
- Manufactured in compliance with ISO 594-1 standards, these fittings are inspected to be free from contaminants, and are fully traceable.
- All parts meet tight dimensional tolerances for increased reliability and process purity.
- Choose from a range of sizes, 1⁄16 to 1⁄4in. with a variety of connection options.
- Sold in packs of 25.
Specifications
Female luer to 1/8 in. low-profile semi-rigid tubing barb | |
Polypropylene | |
25/Pk. |