Learn More
Hamilton™ Female Luer / M6 Threads
Hamilton offers a diverse line of inert fittings fittings and connectors made from CTFE. The fittings are designed for use with industry standard luer and luer lock products like our syringes as well as threaded 1/4-28 and M6 flat bottom ports.
Brand: Hamilton™ 32874
This item is not returnable.
View return policy
Description
Specification Sheet
Connection 1 | Female Luer lock |
---|---|
Connection 2 | Male M6 |
Sales Quantity | 1 each |
I.D. | 1.5 mm |
Specifications
1.5 mm | |
Female Luer lock | |
Each |