Learn More
Value Plastics Female Luer Thread Style to NPT Adapter Fittings, Straight
Shop All Cole-Parmer Products
Click to view available options
Material (Seal):
Prolypropelene
White Nylon
Unit Size:
Pack of 25
Description
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Inlet | 1/8-27 NPT(M) Thread |
| Material (Seal) | Prolypropelene |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion