Learn More
Fenamiphos, Honeywell Fluka™
Brand: Honeywell Fluka R1703-100MG
Additional Details : Weight : 0.10000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : II
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Fenamiphos | |
≤ 100% | |
(RS)-(ethyl 4-methylthio-m-tolyl isopropylphosphoramidate) | |
Acaricides and Insecticides | |
49°C | |
100 mg |
22224-92-6 | |
4752893 | |
303.36 g/mol | |
1.191 g/cm3 | |
Colorless | |
200°C |