Learn More
Fenchlorphos, MedChemExpress
MedChemExpress Fenchlorphos, an organophosphate, is an insecticide. Fenchlorphos is an inhibitor of the enzyme acetylcholinesterase (AChE). Fenchlorphos is able to cause mitochondrial dysfunction.
Brand: Medchem Express HY-B1093
This item is not returnable.
View return policy
Description
Fenchlorphos, an organophosphate, is an insecticide. Fenchlorphos is an inhibitor of the enzyme acetylcholinesterase (AChE). Fenchlorphos is able to cause mitochondrial dysfunction.Specifications
Fenchlorphos | |
White | |
C8H8Cl3O3PS | |
DMSO : 100 mg/mL (310.99 mM; Need ultrasonic) | |
321.55 | |
99.89% | |
Solid |
299-84-3 | |
Research | |
100 mg | |
S=P(OC)(OC)OC1=CC(Cl)=C(Cl)C=C1Cl | |
321.55 | |
Research | |
COVID-19-immunoregulation |