missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Fenofibrate, 98%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 79°C to 82°C |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 25 g |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in ether,soluble in acetone,,soluble in ethyl acetate,soluble in acetonitrile,soluble in methanol,soluble in ethanol |
Formula Weight | 360.83 |
Percent Purity | 98% |
Chemical Name or Material | Fenofibrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'