missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Fenretinide
GREENER_CHOICE

Product Code. 18056052
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18056052

Brand: Tocris Bioscience™ 1396/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic retinoid; antiproliferative and antioxidant

Synthetic retinoid agonist. Antiproliferative, antioxidant and anticancer agent with a long half-life in vivo. Apoptotic effects appear to be mediated by a mechanism distinct from that of 'classical' retinoids.

Deuterated analog also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65646-68-6
Molecular Formula C26H33NO2

Specifications

Quantity 10 mg
Formula Weight 391.55
Purity >98%
Molecular Weight (g/mol) 391.555
Chemical Name or Material Fenretinide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.