missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cylindrospermopsin solution, 10 μg/mL in water, Honeywell Fluka™
Click to view available options
Quantity:
1 mL
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Liquid |
CAS | 143545-90-8 |
Grade | Analytical Standard |
Packaging | Glass Bottle |
Quantity | 1 mL |
For Use With (Application) | Cyanobacterial Toxins |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'