Learn More
BRAND™ FEP Filling Tube
Manufactured from FEP. BRAND™ FEP Filling Tube dispenses accurate volumes directly from drums and bulk refills. For use with Dispensette™III/Organic.
Brand: BRAND™ 704209
Code : NEW
Additional Details : Weight : 0.05000kg
Description
- Manufactured from FEP.
- Dispenses accurate volumes directly from drums and bulk refills.
- For use with Dispensette™III/Organic
Specifications
FEP | |
6.0 mm | |
1.5 m | |
Dispensette™III/Organic |