Learn More
Cytiva FEP Tubing Kit
Can be used as inlet or outlet tubing for AKTA Avant systems
Brand:  Cytiva 28-9809-84
Product Code. 12495710
  • £202.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Inlet tubing for ÅKTA avant 25 or outlet tubing for ÅKTA avant 150.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tubing Kit
1.6 mm
FEP
1/8 in.
1 Pack