missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Taq Buffer with (NH4)2SO4 (10X)
Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native). Available in different compositions and also without detergents.
Brand: Thermo Scientific™ B33
10.87 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native). Available in different compositions and also without detergents.
10X Taq Buffer with (NH4)2SO4 (B33) includes:
- 750 mM Tris-HCl (pH 8.8 at 25°C)
- 200 mM (NH4)2SO4
- 0.1% (v/v) Tween 20
Related Products
- 10X Taq Buffer (10X), with KCl and 15 mM MgCl2 (B16)
- 10X Taq Buffer (10X), with KCl (B38)
- 10X Taq Buffer (10X), with (NH4)2SO4 and 20 mM MgCl2 (B34)
- 10X Taq Buffer (10X), without detergent (B55)
Specifications
• 10X Taq Buffer with (NH4)2SO4, 4 x 1.25 mL Store at -20°C. |
|
Taq DNA Polymerase | |
4 x 1.25 mL | |
Wet or Dry Ice | |
10X |
Liquid | |
Taq Buffer with (NH4)2SO4 | |
PCR | |
Standard PCR | |
4 x 1.25 mL |
For Research Use Only. Not for use in diagnostic procedures.