Learn More
Thermo Scientific™ DNA Gel Loading Dye (6X)
Prepare DNA markers and samples for loading on agarose or polyacrylamide gels with Thermo Scientific™ 6X DNA Loading Dye.
Brand: Thermo Scientific™ R0611
37.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : 88
Additional Details : Weight : 0.00090kg
Description
6X DNA Loading Dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It contains two different dyes (bromophenol blue and xylene cyanol FF) for visual tracking of DNA migration during electrophoresis. The presence of glycerol ensures that the DNA in the ladder and sample forms a layer at the bottom of the well. The EDTA included in the solution binds divalent metal ions and inhibits metal-dependent nucleases.
6X DNA Loading Dye is used for conventional DNA electrophoresis.
Highlights
- Two-color tracking of DNA migration during DNA electrophoresis
- No DNA masking during gel exposure to UV light
- EDTA binds divalent metal ions and inhibits metal dependent nucleases
Specifications
6 X | |
DNA Gel Loading Dye | |
Acrylamide Gels, Agarose Gels |
For Research Use Only. Not for use in diagnostic procedures.