Learn More
Thermo Scientific™ BclI (10 U/μL)
Cut at T^GATCA sites with BclI restriction enzyme, which performs best at 55°C in G buffer (Isoschizomers: FbaI, Ksp22I).
Brand:  Thermo Scientific™ ER0722
 View more versions of this product
Product Code. 10569120
  • £90.00 / 5000 units
Explore more special offers
Description
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality – stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Applications
  • Molecular cloning
  • Restriction site mapping
  • Genotyping
  • Southern Blot
  • Restriction fragment length polymorphism (RFLP)
  • SNP
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 U/μL
BclI
Yes
No
Restriction Enzyme
Dam Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer G
55°C
5,000 units
Traditional Cloning