missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Biotin Chromogenic Detection Kit

Product Code. 10899710 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 Reactions
30 Reactions
Unit Size:
10 reactions
30 reactions
This item is not returnable. View return policy

Product Code. 10899710

Brand: Thermo Scientific™ K0662

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific Biotin Chromogenic Detection Kit is a convenient tool for the chromogenic detection of biotinylated nucleic acid probes.

Thermo Scientific Biotin Chromogenic Detection Kit is a convenient tool for the chromogenic detection of biotinylated nucleic acid probes. The kit is optimized to reproducibly provide high sensitivity with a low background in applications, such as Southern, Northern, dot and slot blotting, and screening of viral plaques and bacterial colonies. Biotinylation of DNA/RNA probes is widely used as a safe and convenient alternative to radioactive labeling. Biotin can be incorporated into nucleic acids using various enzymatic or non-enzymatic methods, including the Biotin DecaLabel DNA Labeling Kit (Cat. No. K0651).

Features
Biotinylated probes are detected with streptavidin coupled to alkaline phosphatase (AP). Streptavidin-AP conjugates bind specifically and irreversibly to the biotin-labeled probes. The probes are then visualized using a chromogenic substrate for alkaline phosphatase BCIP/NBT, which produces a blue-purple precipitate. Visualization does not require X-ray film or other specific equipment (see Figure 1).

Highlights
High sensitivity of detection—0.03 pg of homologous DNA in dot blot hybridization with biotin-labeled probe
Low background due to optimized washing and blocking procedures
Non-radioactive detection visualization does not require X-ray film or other specific equipment (see Figure 2)
Fast and convenient—ready-to-use components

Applications
• Southern, Northern, dot/slot blots, plaque, or colony screening.

Note: This kit is optimized for hybridization procedures using nylon membranes. The detection sensitivity may vary depending on the membranes supplied by different vendors.

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Chromogenic
Format Kit
Labeling Target DNA/RNA
For Use With (Application) Southern, Northern, Dot/Slot Blots, Plaque, Colony Screening
Product Type Biotinylated Nucleic Acid Detection Kit
Quantity 30 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.