Learn More
Thermo Scientific™ BseMI (BsrDI) (5 U/μL)
Cut at GCAATG(2/0)^ sites with BseMI (BsrDI) restriction enzyme, which performs best at 55°C in R buffer (Isoschizomers: Bse3DI, BsrDI).
Brand: Thermo Scientific™ ER1261
17.81 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
- Superior quality -stringent quality control and industry leading manufacturing process
- Convenient color-coded Five Buffer System
- Includes universal Tango buffer for double-digestions
- BSA premixed in reaction buffers
- Wide selection of restriction endonuclease specificities
Specifications
5 U/μL | |
BseMI (BsrDI) | |
Yes | |
Yes | |
Restriction Enzyme |
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
10X Buffer R | |
55°C | |
100 U | |
Scarless Cloning |
For Research Use Only. Not for use in diagnostic procedures.