missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BseNI (BsrI) (10 U/μL)

Product Code. p-4521387 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1,000 units
5000 U
Unit Size:
1000 units
5000 units
This item is not returnable. View return policy

Product Code. 10161270

Brand: Thermo Scientific™ ER0881

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cut at ACTGG(1/-1)^ sites with BseNI (BsrI) restriction enzyme, which performs best at 65°C in B buffer (Isoschizomers: Bse1I, BsrI, BsrSI).

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...A C T G G N▵...3'

3'...T G A C▵C N ...5'

Conditions for 100% Activity:

  • 1X Buffer B:10mM Tris-HCl (pH7.5 at 37°C), 10mM MgCl2 and 0.1mg/mL BSA
  • Incubate at 65°C
  • To ensure higher efficiency of digestion, perform the cleavage reaction under paraffin oil

Storage Buffer:

  • BseNI is supplied in:10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with BseNI, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: never overlaps — no effect
  • EcoKI: may overlap — effect not determined
  • EcoBI: may overlap — effect not determined

Note:

Incubation at 37°C results in less than 10% activity.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme BseNI (BsrI)
Compatible Buffer 10X Buffer B
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 65°C
Type IIS RE Yes
Quantity 1,000 units
Product Type Restriction Enzyme
Research Category Scarless Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.