missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BseXI (BbvI) (3 U/μL)

Cut at GCAGC(8/12)^ sites with BseXI (BbvI) restriction enzyme, which performs best at 65°C in its own unique buffer (Isoschizomers: BbvI, BstV1I).

Brand:  Thermo Scientific™ ER1452

82.00 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.


 View more versions of this product

Product Code. 10714111

  • £123.00 / 500 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price

This item is not returnable. View return policy

Description

Description

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...G C A G C (N)8▵...3'

3'...C G T C G (N)12▵...5'

Conditions for 100% Activity:

  • 1X Buffer BseXI: 50mM Tris-HCl (pH 7.5 at 37°C), 2mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 65°C
  • To ensure higher efficiency of digestion, perform the cleavage reaction under paraffin oil

Storage Buffer:

  • BseXI is supplied in:10mM Tris-HCl (pH 7.4 at 25°C), 200mM NaCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 10-fold overdigestion with BseXI, more than 95% of the DNA fragments can be ligated recut

Methylation Effects:

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: may overlap — no effect
  • EcoKI: never overlaps — no effect
  • EcoBI: may overlap — effect not determined

Note:

Assayed using pBR322 DNA (#SD0041). Incubation at 37°C results in 10% activity. Greater than 40-fold overdigestion with BseXI may result in star activity. BseXI may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3 U/μL
65°C
500 U
BseXI (BbvI
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.