Learn More
Thermo Scientific™ BshTI (AgeI) (10 U/μL)
Cut at A^CCGGT sites with BshTI (AgeI) restriction enzyme, which performs best at 37°C in O buffer (Isoschizomers: AgeI, AsiGI, CspAI, PinAI).
Brand: Thermo Scientific™ ER1461
Code : 88
Additional Details : Weight : 1.51950kg
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...A▵C C G G T...3'
3'...T G G C C▵A...5'
Conditions for 100% Activity:
- 1X Buffer O: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- BshTI is supplied in:10mM Tris-HCl (pH7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Ligation and Recleavage:
- After 50-fold overdigestion with BshTI, more than 95% of the DNA fragments can be ligated and recut
Methylation Effects:
- Dam: never overlaps — no effect
- Dcm: never overlaps — no effect
- CpG: completely overlaps — blocked
- EcoKI: may overlap — effect not determined
- EcoBI: may overlap — effect not determined
Digestion of Agarose-embedded DNA:
- Minimum 20units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours
Note:
Low salt, high glycerol (>5%) concentrations, pH >8.0 or a large excess of enzyme may result in star activity.
Specifications
10 U/μL | |
BshTI (AgeI) | |
Yes | |
No | |
Restriction Enzyme |
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive | |
10X Buffer O | |
37°C | |
200 U | |
Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.