Learn More
Thermo Scientific™ CloneJET PCR Cloning Kit
Efficiently clone PCR products generated with any thermostable DNA polymerase using Thermo Scientific™ CloneJET™ PCR Cloning Kit, a positive selection system.
Brand: Thermo Scientific™ K1232
212.43 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
CloneJET PCR Cloning Kit is an advanced positive selection system for high-efficiency cloning of PCR products generated with any thermostable DNA polymerase. Any other blunt or sticky-end DNA fragment can be cloned. It is ideal for phosphorylated or non-phosphorylated DNA fragments. Ligation into the included positive selection vector takes only 5 minutes, yielding more than 99% recombinant clones. Blunt-ended PCR products generated with a proofreading enzyme are ligated directly into the cloning vector.
PCR products generated either with non-proofreading DNA polymerases or mixtures of DNA polymerases are blunted prior to ligation in 5 minutes with the thermostable DNA Blunting Enzyme provided with the kit. All common laboratory E. coli strains can be directly transformed with the ligation product.
Additionally, any other blunt or sticky-end DNA fragment can be cloned. The kit is ideal for phosphorylated or non-phosphorylated DNA fragments. Ligation into the included positive selection vector takes only 5 minutes and yields more than 99% recombinant clones. Blunt-ended PCR products generated with a proofreading enzyme (e.g., Pfu polymerase) are ligated directly into the cloning vector. PCR products generated either with non-proofreading DNA polymerases (e.g., Taq polymerase) or mixtures of DNA polymerases are blunted prior to ligation in 5 minutes with the thermostable DNA Blunting Enzyme provided with the kit. All common laboratory E.coli strains can be directly transformed with the ligation product.The kit contains a novel, ready-to-use positive selection cloning vector pJET1.2/blunt. The vector contains a lethal restriction enzyme gene that is disrupted by ligation of a DNA insert into the cloning site. As a result, only bacterial cells with recombinant plasmids are able to form colonies. Recircularized pJET1.2/blunt vector molecules lacking an insert express a lethal restriction enzyme which kills the host E.coli cell after transformation. This positive selection drastically accelerates the process of colony screening and eliminates additional costs required for blue/white selection. For convenience in mapping and manipulation of the insert, the pJET1.2/blunt cloning vector multiple cloning site contains two BglII recognition sequences that flank the insertion site. In addition, the vector contains a T7 promoter for in vitro and in vivo transcription as well as sequencing of the insert.
- Fast—PCR cloning in only 5 minutes
- Highest efficiency—>99% of positive clones
- No cloning background—positive selection vector
- Versatile—ideal for blunt-end or sticky-end cloning
- Economical—no expensive blue/white screening
Recommended for:
Cloning of blunt-end or 3'-dA tailed PCR products up to 10kb; Cloning of DNA fragments generated by restriction enzymes; Sequencing of cloned DNA; In vitro and in vivo transcription of cloned inserts from the T7 promoter
Note:
Prior to electroporation always column-purify the ligation mixture using, e.g., GeneJET PCR Purification Kit, †K0701, or chloroform extract kit. Electroporation is inhibited by the presence of proteins and salts in the mixture.
Specifications
Ampicillin (AmpR) | |
Kit | |
PCR Cloning | |
CloneJET | |
40 Reactions |
Restriction Enzyme⁄ MCS, Blunt PCR | |
PCR Products | |
40 Reactions | |
Cloning Kit | |
pJET1.2/blunt |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.