missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CTP Solution (100 mM)

Product Code. 10839540 Shop All Thermo Scientific Products
Click to view available options
Quantity:
0.25 mL
Unit Size:
0.25mL
This item is not returnable. View return policy

Product Code. 10839540

Brand: Thermo Scientific™ R0451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific CTP (cytidine 5'-triphosphate) is an extremely stable nucleotide with more than 99% purity.

Thermo Scientific CTP (cytidine 5'-triphosphate) is an extremely stable nucleotide with more than 99% purity. It is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. This nucleotide is designed for a variety of molecular biology applications.

Features
• Greater than 99% purity confirmed by HPLC
• Functionally tested in in vitro transcription
• Stable for two years at -20°C and after multiple freeze-thaw cycles

Applications
• aRNA synthesis
• siRNA synthesis
• RNA amplification
in vitro transcription

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100 mM
For Use With (Application) in vitro Transcription, RNA Amplification
Content And Storage CTP Solution, 100 mM (0.25 mL)

Store at –20°C.
Shipping Condition Dry Ice
Conjugate Unconjugated
Quantity 0.25 mL
Product Type CTP Solution
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.