Learn More
Thermo Scientific™ Biotin DecaLabel DNA Labeling Kit
Efficiently synthesize biotin-labeled DNA probes with the Thermo Scientific™ Biotin DecaLabel DNA Labeling Kit.
£117.00 - £290.00
Specifications
Labeling Target | Genomic DNA, PCR Products, DNA (General) |
---|---|
No. of Reactions | 10 |
Quantity | 10 Reactions |
Description
Biotin DecaLabel DNA Labeling Kit is an advanced system for the efficient synthesis of biotin-labeled DNA probes, based on an improved random-primed labeling method developed by Feinberg and Vogelstein. The primary improvement over traditional random-primed method involves the use of random decamers instead of hexamers, ensuring more efficient annealing with DNA at 37°C. Klenow Fragment, exo-, included in the kit, is genetically engineered enzyme with no detectable exonuclease activity. The enzyme does not degrade the labeled probe during reaction, which results in a high labeling yield even with low amounts of template. You can uniformly label any length DNA fragments.
Biotin-labeled DNA is detected with the Biotin Chromogenic Detection Kit or conventional biotin-avidin or biotin-streptavidin detection systems.
Highlights
- Non-radioactive labeling of DNA
- Efficient priming of labeling reactions with random decamers
- High yields with Klenow Fragment, exo-: no degradation of a labeled probe during reaction
Specifications
Genomic DNA, PCR Products, DNA (General) | |
10 | |
10 Reactions |