missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dilution Buffer for Restriction Enzymes

Optimize applications that require diluted enzymes with this dilution buffer.

Brand:  Thermo Scientific™ B19

Product Code. 11889602

  • £15.67 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Dilution Buffer for Restriction Enzymes is used for applications that require diluted enzymes. Enzymes diluted in this buffer retain 50 to 100% activity after storage for one month at -20°C.

Composition:
  • 10 mM Tris-HCl (pH 7.4 at 25°C)
  • 100 mM KCl
  • 1 mM EDTA
  • 1 mM DTT
  • 50% glycerol
  • 0.2 mg/mL BSA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 x 1.0 mL
Dilution Buffer
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.