Learn More
Thermo Scientific™ DreamTaq Green DNA Polymerase (5 U/μL)
Save time in standard PCR applications with this enhanced Taq DNA polymerase, supplied with colored buffer enabling direct gel loading.
£39.52 - £1126.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10689320
|
Thermo Scientific™
EP0711 |
200U (5U/μL) |
£39.52
200 units |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10776411
|
Thermo Scientific™
EP0712 |
500U (5U/μL) |
£75.25
500 units |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11803823
|
Thermo Scientific™
EP0713 |
5 x 500U (5U/μL) |
£323.00
Pack of 5 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10262450
|
Thermo Scientific™
EP0714 |
20 x 500U (5U/μL) |
£1126.00
Pack of 20 |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15757496
|
Thermo Scientific™
EP0713 |
5 x 500U (5U/μL) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15747496
|
Thermo Scientific™
EP0712 |
500U (5U/μL) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
DreamTaq Green DNA Polymerase is a combination of DreamTaq DNA Polymerase and 10X DreamTaq Green Buffer. DreamTaq DNA Polymerase is an enhanced Taq polymerase optimized for high throughput PCR applications. It ensures higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required.
The 10X DreamTaq Green Buffer includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with PCR performance and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion. For applications requiring PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless 10X DreamTaq Buffer (#B65) or purifying the PCR product before analysis. The 10X DreamTaq Green Buffer is optimized for robust performance in PCR and includes MgCl2 at a concentration of 20 mM.
DreamTaq Green PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq Green buffer, MgCl2, and dNTPs. The master mix retains all features of DreamTaq DNA Polymerase. It is capable of robust amplification of up to 6 kb from genomic DNA and up to 20 kb from viral DNA.
Highlights
- Direct loading of PCR product on gel
- High yields and high sensitivity of PCR
- Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
- Robust amplification with minimal optimization
- Incorporates modified nucleotides, but does not incorporate dUTP
For Research Use Only. Not for use in diagnostic procedures.