Learn More
Thermo Scientific™ DTT (dithiothreitol)
Stabilize enzymes and other proteins which possess free sulfhydryl groups with DTT, shown to restore activity lost by oxidation of these groups in vitro.
£50.75 - £160.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10699530
|
Thermo Scientific™
R0861 |
5 g |
£50.75
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10578170
|
Thermo Scientific™
R0862 |
25 g |
£160.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DTT (DL-Dithiothreitol; Clelands reagent) is used to stabilize enzymes and other proteins which possess free sulfhydryl groups. It has been shown to restore activity lost by oxidation of these groups in vitro. DTT quantitatively reduces disulfide bonds and maintains monothiols in a reduced state .
At a final 0.1 M concentration, DTT is also widely used for disruption of protein disulfide bonds in SDS-polyacrylamide gel electrophoresis.
Highlights
- Free of endo-, exodeoxyribonucleases, ribonucleases, and phosphatases.
Specifications
0.1 M | |
DTT (Dithiothreitol) |
Safety and Handling
- DTT
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
Supplemental information
- MIXTURE LIST-Contains : 2, 3-Butanediol, 1,4-dimercapto-, (R*,R*)-
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.