Learn More
Thermo Scientific™ Fast DNA End Repair Kit
Perform blunting and phosphorylation of DNA ends in just 5 minutes for subsequent use in blunt-end ligation with this kit.
Brand: Thermo Scientific™ K0771
132.00 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : A0
Additional Details : Weight : 1.51950kg
Description
Fast DNA End Repair Kit is used for blunting and phosphorylation of DNA ends in just 5 minutes for subsequent use in blunt-end ligation.
All components of the kit contain premixed reagents to reduce pipetting steps and provide convenience. The End Repair Enzyme Mix contains an optimized mixture of T4 DNA Polymerase and Klenow Fragment to achieve highly effective blunting of fragmented DNA, and T4 Polynucleotide Kinase (PKN) for efficient phosphorylation of DNA ends. The 10X End Repair Reaction Mix contains an optimized reaction buffer, ATP, and dNTPs.
Samples such as fragmented genomic DNA (restriction enzyme digested, nebulized or sonicated), restriction enzyme digested plasmid DNA, double stranded cDNA, and PCR products containing dA overhangs are all compatible with the kit.
Highlights
During the DNA end repair reaction, fragmented DNA is converted into blunt-end DNA containing a 5'-phosphate and 3'-hydroxyl groups. The 5'→3' polymerase activity of the End Repair Enzyme Mix fills-in 5' protruded DNA ends while 3'→5' exonuclease activity removes 3'-overhangs. T4 PNK adds 5'-phosphates to ends of unphosphorylated DNA fragments, such as PCR products.
Highlights
- Efficient - blunting and phosphorylation of 0.5 to 5 µg DNA
- Fast -reaction is completed in 5 minutes
- Convenient -reaction components are premixed to reduce pipetting steps
Specifications
Kit | |
DNA Polymerase, T4 Polynucleotide Kinase | |
50 Reactions | |
50 Reactions |
DNA | |
Cloning | |
Fast DNA End Repair Kit |
For Research Use Only. Not for use in diagnostic procedures.