Learn More
Thermo Scientific™ GeneJET PCR Purification Kit
Thermo Scientific GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.
Brand: Thermo Scientific™ K0701
58.22 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Thermo Scientific GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions. It effectively removes primers, dNTPs, unincorporated labeled nucleotides, enzymes, and salts from PCR and other reaction mixtures.
The kit can be used for purification of DNA fragments from 25 bp to 20 kb with recovery rates up to 100%. Each GeneJET purification column has a total binding capacity of up to 25 μg of DNA, and the entire procedure takes 5 minutes. The purified DNA can be used in common downstream applications, such as sequencing, restriction digestion, labeling, ligation, cloning, in vitro transcription, blotting or in situ hybridization.
Highlights
• Highly efficient: 90 to 100% recoveries in the range of 100 bp to 10 kb
• Fast: procedure takes only 5 minutes
• Convenient: spin columns are capped and assembled with collection tubes
Applications
Fast and efficient purification of DNA fragments ideal for use in all conventional molecular biology procedures including:
• FastDigest or conventional restriction digestion
• Automated fluorescent or radioactive sequencing
• PCR
• In vitro transcription
• In situ hybridization
• Labeling
• Cloning
Specifications
Spin Column | |
GeneJET | |
50 Preps | |
DNA from Enzymatic Reactions (e.g. PCR) | |
Spin Column |
Not High-throughput Compatible (Manual) | |
PCR Purification Kit | |
5 min. | |
Real Time PCR (qPCR), Cloning, DNA Sequencing | |
dsDNA |
Safety and Handling
- genejet PCR purif
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : thiocyanic acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.